Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
/
Northern Ireland
Products
Services
Digital Solutions
Applications
Support
About
£0.00
Order Entry
Home
Wesson salt mixture
Wesson salt mixture, powder
Wesson salt mixture, powder
Wesson salt mixture, powder
:
MP Biomedicals
Wesson salt mixture, powder
:
MP Biomedicals
:
Environmentally Preferable:
Formulation:
Consistent with Fomula, see Product Datasheet for full formulation
Form:
Powder
Storage:
Store at Room Temperature (15 - 30 °C)
Pk
ICNA0290285102
0290285102
2 kg
1 * 2 kg
1 * 2 kg - £0.00
£0.00
-
+
1 * 2 kg
£0.00
-
+
ICNA0290285110
0290285110
10 kg
1 * 10 kg
1 * 10 kg - £344.00
£344.00
-
+
1 * 10 kg
£344.00
-
+