Immersed body (Plummet)
Supplier: Industrial Physics
SIMEDV5000EA 891 GBP
SIMEDV5000
Immersed body (Plummet)
The immersed body (260 mm) is designed to determine the density of paints or related products at 23 °C.
The method is based on 'Archimedes' principle.
The upward force caused by immersing the plummet into the liquid is read from a laboratory balance and expressed in grams per milliliter.
Certifications: DIN EN ISO 2811-2
Delivery information: Supplied in plastic case - A stand needs to be ordered separately.