Crucibles with cover plate
44595.KTEA 273 GBP
ALFA44595.KT
Crucibles with cover plate
These crucibles are manufactured from magnesium aluminium oxide.
  • Superior characteristics at elevated temperature and at high frequencies
  • Resistant to corrosive liquids and gases
  • Inert to oxidation
  • Not subject to radiation damage