Filter capsules with Posidyne® membrane, Kleenpak™
Supplier: Pall Laboratory
PALSKA2NTZP3EA 0 GBP
PALSKA3NFZP1S PALSKA2NTZP3 PALSKA3NFZP6S PALSKA1NFZP2S PALSKA4NFZP6S
Filter capsules with Posidyne® membrane, Kleenpak™
Nylon 6,6 positively charged membrane with EPDM vent and PE support. These membrane are rugged, self-contained sanitary filters designed for small-batch sterile filtration of aqueous pharmaceutical solutions. It can be effective in removing contaminants such as cell debris and endotoxin smaller than the filter rating. It is suitable for high protein recovery from serum and most protein solutions.
  • Net positive charge ranges from pH 3 to 10
  • Double-layer sterilising membranes for high reliability
  • Sanitary vent and drain valves with non-twist hose barbs
Certifications: Meets USP biological reactivity test, in vivo, for Class VI-121 °C Plastics and effluent quality test.