Newborn bovine calf serum, iron-supplemented, New Zealand Origin, HyClone™
Supplier: HyClone products (Cytiva)
HYCLSH30626.02EA 94.5 GBP
HYCLSH30626.02 HYCLSH30626.03 HYCLSH30626.01IR HYCLSH30626.01 HYCLSH30626.03HI HYCLSH30626.02HI
Newborn bovine calf serum, iron-supplemented, New Zealand Origin, HyClone™
Newborn bovine calf serum (NCS), iron supplemented, contains three to four times as much available iron and transferrin as FBS or equine serum. The serum is sourced from animals that are typically less than 10 days old and sterile filtered using sequential 100 nm (0,1 μm) pore-size rated filters.