Northern Ireland
Order Entry
1H-Benzimidazole-5-boronic acid pinacol ester 97%
 
1H-Benzimidazole-5-boronic acid pinacol ester 97%
Catalog # ACRO430540010
CAS Number:  
undefined
 
1H-Benzimidazole-5-boronic acid pinacol ester 97%
Catalog # ACRO430540010
CAS Number:  
 
£262.00
   
  1 * 1 g

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to tan to purple Crystalline powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Titration with NaOH:
    ≥96.0 %

Specification Test Results

About this item

 :
 :244.10 g/mol
 :204,2…208.5 °C
 :GHS07
 :Ambient
 :1007206-54-3
 :C₁₃H₁₇BN₂O₂
 :MFCD11054041