
Tabs 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54960-65-5
:
Formula:C₈H₁₉NO₆S
MDL Number:MFCD00678129
:257.301 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Solubility:(50 % in water) Clear colorless
- Titration with NaOH:≥98.5 % (On anhydrous basis)
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: 428080250Local Article Number: ACRO428080250
- Supplier Article Number: 428081000Local Article Number: ACRO428081000