(S)-(+)-1-Boc-3-hydroxypyrrolidine 98+%
ACRO397580050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to light yellow Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- HPLC:≥98.0 %
- Specific optical rotation:+21° to +26° (20°C, 589 nm) (C=1, in Methanol)
:101469-92-5
:
:C₉H₁₇NO₃
:MFCD01317839
:187.2362 g/mol
:60…64 °C
:GHS07
:Refrigerator