Northern Ireland
Order Entry
Griess' reagent
Griess' reagent
  ACRO328670500
 :  
undefined
Griess' reagent
  ACRO328670500
 :  
 
£134.00
   
  1 * 50 mL

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370
  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless to light pink Solution
  • Suitability:
    (for NO2 det.in biol.media) Passes test
  • Composition:
    0.1% naphthylethylenediamine dihydrochloride

 

 

 :
 :1,03 g/cm³
 :GHS07
 :Refrigerator
 :MFCD00240366