Northern Ireland
Order Entry
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
 :  Biotium
 
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
 :  Biotium

 

  • Catalog No:
  • 20418-50UL
  • 20418-250UL
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Rabbit IgG
  • Rabbit IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 680
  • CF® 680
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Donkey
  • Donkey
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • Cross adsorption:
  • Bovine, chicken, goat, guinea pig, horse, human, mouse, sheep, Syrian hamster
  • Bovine, chicken, goat, guinea pig, horse, human, mouse, sheep, Syrian hamster
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.

 

 

  •  
     
    Size
    Pk
     
     
  • 20418-50UL
    20418-50UL
    50 uL
    50 µl
       
       
    1 * 50 µl
    1 * 50 µl - £145.00
    £145.00
    1 * 50 µl
       £145.00

     
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Rabbit IgG
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    CF® 680
    Reactivity
    Rabbit
    Host
    Donkey
    Isotype
    Form
    Liquid
    Purification
    Affinity purified
    Concentration
    2 mg/mL
    Cross adsorption
    Bovine, chicken, goat, guinea pig, horse, human, mouse, sheep, Syrian hamster
    Storage buffer
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
    Storage temperature
    Store at –20 °C.
  • 20418-250UL
    20418-250UL
    250 μL
    250 µl
       
       
    1 * 250 µl
    1 * 250 µl - £346.00
    £346.00
    1 * 250 µl
       £346.00

     
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Rabbit IgG
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    CF® 680
    Reactivity
    Rabbit
    Host
    Donkey
    Isotype
    Form
    Liquid
    Purification
    Affinity purified
    Concentration
    2 mg/mL
    Cross adsorption
    Bovine, chicken, goat, guinea pig, horse, human, mouse, sheep, Syrian hamster
    Storage buffer
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
    Storage temperature
    Store at –20 °C.