This product is prepared by labeling highly cross-adsorbed donkey anti-goat IgG (H+L) with CF™405S. CF™405S is a blue fluorescent dye with an absorption peak wavelength that nearly coincides with the 405 nm blue diode laser line. In addition, the emission peak wavelength of the dye well centers within the blue detection window of BD flow cytometers. As a result, flow cytometry analysis using CF™405S dye results in much brighter signal than that using Alexa Fluor® 405. To minimize cross reactivity, the antibody has been adsorbed against chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit, and rat serum proteins.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF405S
Epitope:
Host: Donkey
Isotype:
Reactivity: Goat
- Catalog No:
- 20416-500UL
- 20416-50UL
- Antigen symbol:
- IgG
- IgG
- Antigen name:
- Goat IgG
- Goat IgG
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Reactivity:
- Goat
- Goat
- Host:
- Donkey
- Donkey
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Concentration:
- 2 mg/mL
- 2 mg/mL
- Cross adsorption:
- Chicken, guinea pig, horse, human, mouse, rabbit, rat, Syrian hamster
- Chicken, guinea pig, horse, human, mouse, rabbit, rat, Syrian hamster
- Storage buffer:
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:
- Store at –20 °C.
- Store at –20 °C.
- SizePk
Specifications
Antigen symbolIgGAntigen nameGoat IgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationCF® 405SReactivityGoatHostDonkeyIsotypeFormLiquidPurificationAffinity purifiedConcentration2 mg/mLCross adsorptionChicken, guinea pig, horse, human, mouse, rabbit, rat, Syrian hamsterStorage bufferPBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azideStorage temperatureStore at –20 °C.Specifications
Antigen symbolIgGAntigen nameGoat IgGEnvironmentally PreferableAntibody typeSecondaryClonalityPolyclonalConjugationCF® 405SReactivityGoatHostDonkeyIsotypeFormLiquidPurificationAffinity purifiedConcentration2 mg/mLCross adsorptionChicken, guinea pig, horse, human, mouse, rabbit, rat, Syrian hamsterStorage bufferPBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azideStorage temperatureStore at –20 °C.