Anti-ICOSLG Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB12839EA 317 GBP
EVBIEB12839
Anti-ICOSLG Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 33kDa band observed in Human Lymph Node lysates (calculated MW of 33.3kDa according to NP_056074.1). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: ICOSLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR