Anti-HLA-B Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB12828EA 317 GBP
EVBIEB12828
Anti-HLA-B Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 40+45kDa bands observed in lysates of cell line Jurkat (calculated MW of 40.5kDa according to NP_005505.2). The observed 45kDa band corresponds to the glycosylated form.Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: HLA-B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR