Anti-SERPINF1 Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB12223EA 317 GBP
EVBIEB12223
Anti-SERPINF1 Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 48kDa band observed in Human Albumin-depleted Serum and Plasma lysates (calculated MW of 46.3kDa according to NP_002606.3). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: SERPINF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Dog, Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR