Anti-ICAM1 Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB11754EA 248 GBP
EVBIEB11754
Anti-ICAM1 Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 60kDa band observed in lysates of cell lines Daudi and HeLa (calculated MW of 57.8kDa according to NP_000192.2). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR