Anti-RAB9A Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9884EA 563 GBP
BSBTPB9884
Anti-RAB9A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR