Back pressure regulator
Supplier: Perkin Elmer
PERC02540272EA 0 GBP
PERC02540272
Back pressure regulator
This back pressure regulator can be used to generate backpressure for applications less than 250 psi.