Anti-XAB2 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP17545CEA 339 GBP
ABGEAP17545C
Anti-XAB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XAB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR