Anti-EDEM3 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP16556BEA 313 GBP
ABGEAP16556B
Anti-EDEM3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EDEM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR