Anti-CPPED1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP17964BEA 313 GBP
ABGEAP17964B
Anti-CPPED1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPPED1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR