Anti-HDLBP Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-6339REA 648 GBP
BOSSBS-6339R
Anti-HDLBP Rabbit Polyclonal Antibody
Antibodies
High density lipoprotein-binding protein, HDLBP, also known as vigilin, specifically binds HDL molecules and may function in the removal of excess cellular cholesterol. It may also be involved in the regulation of the delivery of fats to cells for energy and storage as fats in the bloodstream are not readily metabolized in the absence of HDLBP.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HDLBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR