Anti-TAS2R20 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-11789REA 326 GBP
BOSSBS-11789R
Anti-TAS2R20 Rabbit Polyclonal Antibody
Antibodies
The sense of taste provides animals with valuable information about the quality and nutritional value of food. A family of G protein coupled receptors are involved in taste perception and include T1R, which is involved in sweet and umami taste perception and T2R, which is involved in bitter taste perception. Both types of taste receptors couple to various G proteins to initiate signal transduction cascades. T2R49 plays a role in sensing the chemical composition of the gastrointestinal content. T2R49 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells.

  • WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: TAS2R20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR