Anti-KDM1 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-10166REA 648 GBP
BOSSBS-10166R
Anti-KDM1 Rabbit Polyclonal Antibody
Antibodies
Gene activation and repression is specifically regulated by histone methylation status at distinct lysine residues. Lysine specific demethylase 1 (KDM1/LSD1) is a long-sought histone demethylase that specifically demethylates mono and di methyl histone H3 at K4 and K9. Thus KDM1 is a specific tag for epigenetic transcriptional activation, thereby acting as a corepressor.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: KDM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR