Distilling receiver acc. to Dean-Stark, KIMBLE®
Supplier: DWK Life Sciences
KIMAX®
SCERSP22012-10EA 387 GBP
SCERSP22012-10
Distilling receiver acc. to Dean-Stark, KIMBLE®
Dean stark distilling receiver, made of KIMAX® borosilicate glass, with full length ST joints and a siphon return arm.
  • Very good chemical and temperature resistance
  • Sidearm bridge equalizes pressure
  • Designed from ASTM Specification E123, Style A
Capacity: 10 ml, length: 237 mm.