Would you like to log in again?
Plasmid Miniprep Kit II, peqGOLD
Supplier: VWR Peqlab
peqGOLD
732-3103EA 258 GBP InStock
732-3102 732-3101 732-3103
Plasmid Miniprep Kit II, peqGOLD
Nucleic Acid Purification Systems and Kits Plasmid Purification Systems and Kits
Plasmid Miniprep Kit II for the easy, quick and efficient extraction of highly pure plasmid DNA from up to 15 ml culture. Yields of up to 75 µg high quality plasmid DNA per preparation. Plasmid DNA obtained with peqGOLD Plasmid Miniprep kits can directly be used for a wide range of downstream applications.
  • Suitable for the simultaneous processing of multiple samples
  • Short hands-on and preparation times (<30 min)
  • No need for organic extractions or precipitations
  • Complete enzymatic degradation of all cellular RNA
  • Efficient removal of enzyme inhibitors and contaminants
  • Final elution of DNA with Elution Buffer or water
*Classic Line columns without lids for easy handling.
Delivery information: Each kit includes PerfectBind DNA columns (Safety or Classic Line*) and collection tubes, RNase A Solution, Solution I, II and III, PW Plasmid and DNA Wash Buffer, and Elution Buffer.
Order Now
ORDER
ACCESSORIES
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty