Lysine acetylation of histones and non-histone proteins plays an important part in many cellular processes such as chromatin and nuclear signaling, transcription, gene silencing, cell cycle progression, apoptosis, differentiation, DNA replication and repair.
The antibody specifically binds to proteins with acetylated lysine residues. The antibody has been testeed in western blot and ELISA and is expected to also work in immunoprecipitation if a rabbit-anti-chicken IgG or other IgY binding capture unit is used, however IP is currently untested for this antibody.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Acetyl Lysine (AcK)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Many Species