Anti-Filamin A Mouse Monoclonal Antibody [clone: 4E10-1B2]

Supplier: Biosensis
BSENM-835-100EA 315 GBP
BSENM-835-100
Anti-Filamin A Mouse Monoclonal Antibody [clone: 4E10-1B2]
Antibodies
Filamin A, is a 280-kD protein that promotes orthogonal branching of actin filaments cortical cytoplasm. It anchors various  transmembrane proteins to the actin cytoskeleton and serves as a scaffold for a wide range of cytoplasmic signaling proteins. Remodeling of the cytoskeleton is central to the modulation of cell shape and migration.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, IHC and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Filamin A
Clonality: Monoclonal
Clone: 4E10-1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR