Glycine receptor is a neurotransmitter-gated ion channel. When bound to its ligand, glycine, chloride conductance is increased and consequently inhibits neuronal firing. It is a pentameric receptor composed of alpha and beta subunits and localises to the cell junction, synapse and postsynaptic cell membrane. Defects in GLRA1 cause startle disease (STHE) which is also known as hereditary hyperekplexia or congenital stiff-person syndrome.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: glycine receptor subunit alpha-1 ]
Clonality: Monoclonal
Clone: 2E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human