Orexin receptor type 2 is a nonselective, high-affinity receptor for both orexin-A and orexin-B neuropeptides. It is involved in the central feedback mechanism that regulates feeding behaviour. It is a transmembrane protein and belongs to the G-protein coupled receptor 1 family.
Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Orexin receptor type 2
Clonality: Monoclonal
Clone: 1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human