Anti-Semaphorin-7A Mouse Monoclonal Antibody [clone: 3D3]

BSENM-875-100EA 315 GBP
BSENM-875-100
Anti-Semaphorin-7A Mouse Monoclonal Antibody [clone: 3D3]
Semaphorin-7A is a membrane-bound semaphorin that associates with cell surfaces via a glycosylphosphatidylinositol (GPI) linkage. It is an 80-kDa glycoprotein expressed on activated lymphocytes and erythrocytes. It may play an important role in the nervous system and in modulating immune function. It is expressed in placenta, testis, ovary, spleen, brain, spinal cord, lung, heart, adrenal gland, lymph nodes, thymus, intestine and kidney.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Semaphorin-7A
Clonality: Monoclonal
Clone: 3D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR