Anti-SEMA4B Mouse Monoclonal Antibody [clone: 4B2]

BSENM-889-100EA 315 GBP
BSENM-889-100
Anti-SEMA4B Mouse Monoclonal Antibody [clone: 4B2]
SEMA4B is a 837 amino acid polypeptide that has an estimated molecular weight of 92.7 kDa. It contains a Sema domain which occurs in semaphorins, which are a large family of secreted and transmembrane proteins, some of which function as repellent signals during axon guidance. It also contains a a PSI domain (plexin repeat). This is a cysteine rich repeat found in several different extracellular receptors.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SEMA4B
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR