Anti-Teneurin-1 Mouse Monoclonal Antibody [clone: 3G8]

BSENM-884-100EA 315 GBP
BSENM-884-100
Anti-Teneurin-1 Mouse Monoclonal Antibody [clone: 3G8]
Teneurin-1 may function as a cellular signal transducer. It exists as a disulfide-linked homodimer transmembrane protein. It is expressed in fetal brain.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB, and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Teneurin-1
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR