Anti-HESR1 Mouse Monoclonal Antibody [clone: 1E10]

Supplier: Biosensis
BSENM-893-100EA 315 GBP
BSENM-893-100
Anti-HESR1 Mouse Monoclonal Antibody [clone: 1E10]
Antibodies
HESR-1 belongs to the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcriptional repressors. It is a downstream effector of Notch signaling which may be required for cardiovascular development. It binds preferentially to the canonical E box sequence 5'-CACGTG-3'. It represses transcription by the the cardiac transcriptional activators GATA4 and GATA6. HESR1 is expressed in the somitic mesoderm, the central nervous system, the kidney, the heart, nasal epithelium, and limbs. 

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and sandwich ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HESR1
Clonality: Monoclonal
Clone: 1E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR