Anti-Slit-2 Mouse Polyclonal Antibody

Supplier: Biosensis
BSENMO-940-50EA 315 GBP
BSENMO-940-50
Anti-Slit-2 Mouse Polyclonal Antibody
Antibodies
Slit-2 is thought to act as molecular guidance cue in cellular migration, and its function appears to be mediated by interaction with roundabout homolog (ROBO) receptors. It is a secreted protein known for its role of repulsion in axon guidance and neuronal migration that can also inhibit leukocyte chemotaxis induced by chemotactic factors.

Specificity has been confirmed by WB and direct ELISA against the antigen.
Application Information:
This antibody is recommended for WB and ELISA. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Slit-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Slit-2 (1361-1459)
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR