Anti-Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH Rabbit Polyclonal Antibody

Supplier: Biosensis
BSENR-1701-100EA 286 GBP
BSENR-1701-100
Anti-Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH Rabbit Polyclonal Antibody
Antibodies
Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. GAPDH may have other roles in the activation of transcription and in the regulation of apoptosis as well as Alzheimer's disease and Huntington's disease. The immunogen used to raise this particular antibody was extensively purified pig GAPDH. This antibody can be used as a loading control for western blotting experiments, allowing comparison between the level of this protein and others in a cell or tissue.

Specific for the ~36-38kDa Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) protein.
Application Information:
Western Blotting (WB) and Immunocytochemistry (ICC). A dilution of 1:10,000-1:30,000 is recommended for WB. Human GAPDH has a predicted length of 335 residues and a MW of 36 kDa. A dilution of 1:1,000-1:10,000 is recommended for ICC. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Glyceraldehyde 3-Phosphate Dehydrogenase/GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR