HPLC columns, LiChroCART®, with ChiraSep® 100 DNBPG sorbents
MERC1.50236.7005 MERC1.50175.7005 MERC1.51444.7005 MERC1.50201.7005 MERC1.51431.7005 MERC1.50229.7005 MERC1.50233.7005 MERC1.50178.7005 MERC1.50190.7005 MERC1.51448.7005 MERC1.50171.7005 MERC1.51442.7005 MERC1.50195.7005 MERC1.51445.7005 MERC1.50179.7005 MERC1.50234.7005 MERC1.50228.7005 MERC1.50302.7005 MERC1.51443.7005 MERC1.51449.7005 MERC1.51432.7005 MERC1.50170.7005 MERC1.50177.7005 MERC1.51446.7005
HPLC columns, LiChroCART®, with ChiraSep® 100 DNBPG sorbents
Stainless steel LiChroCART® cartridges are available with ChiraSep® sorbents. Ideal for separation of enantiomers with π-electrons.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a 4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of 3 cartridges without cartridge holder. The separate part numbers for the cartridge are as follows: 1.50227.0001 LiChroCART® cartridge holder for 30 mm cartridge and 1.50226.0001 LiChroCART® cartridge holder for 55 mm cartridge.