Anti-M13 Monoclonal Antibody [clone: RL-ph2]

Supplier: Abnova
ABNOMAB1948EA 547 GBP
ABNOMAB1948
Anti-M13 Monoclonal Antibody [clone: RL-ph2]
Mouse monoclonal antibody raised against native M13 phage coat proteins.

Recommended Dilutions: Western Blot: 1:100 to 1:1000; Flow Cytometry: 1:25 to 1:200

Type: Primary
Antigen: M13
Clonality: Monoclonal
Clone: RL-ph2
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR