Anti-CEA Mouse Monoclonal Antibody [clone: 057-10007]

Supplier: Abnova
ABNOMAB4034EA 524 GBP
ABNOMAB4034
Anti-CEA Mouse Monoclonal Antibody [clone: 057-10007]
Mouse monoclonal antibody raised against native Carcinoembryonic antigen.

Type: Primary
Antigen: CEA
Clonality: Monoclonal
Clone: 057-10007
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR