Anti-IP6K1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP16424AEA 313 GBP
ABGEAP16424A
Anti-IP6K1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IP6K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR