Anti-TESK1 Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP16795CEA 339 GBP
ABGEAP16795C
Anti-TESK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TESK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR