Anti-WARS Rabbit Polyclonal Antibody

Supplier: Abgent
ABGEAP18122BEA 313 GBP
ABGEAP18122B
Anti-WARS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: WARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR