Anti-IRAK-1 Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
CAYM13843-1EA 521 GBP
CAYM13843-1
Anti-IRAK-1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRAK-1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR