Anti-Akt1 Mouse Monoclonal Antibody [clone: 104A282]

Supplier: Cayman Chemical
CAYM13733-1EA 484 GBP
CAYM13733-1
Anti-Akt1 Mouse Monoclonal Antibody [clone: 104A282]
Antibodies
Type: Primary
Antigen: Akt1
Clonality: Monoclonal
Clone: 104A282
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR