Anti-Nav1.7 Mouse Monoclonal Antibody [clone: S68-6]

Supplier: Cayman Chemical
CAYM13718-100EA 534 GBP
CAYM13718-100
Anti-Nav1.7 Mouse Monoclonal Antibody [clone: S68-6]
Antibodies
Type: Primary
Antigen: Nav1.7
Clonality: Monoclonal
Clone: S68-6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR