Anti-RORC Rabbit Polyclonal Antibody

Supplier: Cayman Chemical
CAYM11003-1EA 371 GBP
CAYM11003-1
Anti-RORC Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RORC
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Chimpanzee, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR