Anti-ANGPTL3 Rat Monoclonal Antibody [clone: Kairos3-3741]

Supplier: Cayman Chemical
CAYM10807-100EA 435 GBP
CAYM10807-100 CAYM10807-50
Anti-ANGPTL3 Rat Monoclonal Antibody [clone: Kairos3-3741]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: Kairos3-3741
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR