Anti-IL1RN Rabbit Polyclonal Antibody

Supplier: United States Biological
USBI143610EA 398 GBP
USBI143610
Anti-IL1RN Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-2?g/ml

Type: Primary
Antigen: IL1RN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR