Anti-c-erbB2 Mouse Monoclonal Antibody [clone: 3G28]

Supplier: United States Biological
USBIC0026-11V1-250EA 303 GBP
USBIC0026-11V1-250
Anti-c-erbB2 Mouse Monoclonal Antibody [clone: 3G28]
Antibodies
Type: Primary
Antigen: c-erbB2
Clonality: Monoclonal
Clone: 3G28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR