Anti-N-Acetylglucosamine Mouse Monoclonal Antibody [clone: 6D91]

Supplier: United States Biological
USBIA0528-18EA 641 GBP
USBIA0528-18
Anti-N-Acetylglucosamine Mouse Monoclonal Antibody [clone: 6D91]
Antibodies
Recommended dilution: ELISA: 01-1 ?g/ml; Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: N-Acetylglucosamine
Clonality: Monoclonal
Clone: 6D91
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR