Anti-BSA Chicken Polyclonal Antibody

AGRIIMS02-035-326EA 327 GBP
AGRIIMS02-035-326
Anti-BSA Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BSA
Clonality:
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Chicken
Isotype:
Reactivity: A. toxicaria, A. thaliana, d. salina, H. vulgare, P. silvestris, Pisum sativum, S. lycopersicum, Silene vulgaris, Solanum lycopersicum, Solanum tuberosum, Zea mays
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR